Lithium tetrafluoroborate, anhydrous 98%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC22377-0100
AC22377-0500
TS22377-0100EA
56
USD
TS22377-0100
TS22377-0500
Lithium tetrafluoroborate, anhydrous 98%, pure
Lithium tetrafluoroborate
Formula:
BF₄Li MW: 93.74 g/mol Melting Pt: 275 °C (dec.) Density: 0.852 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011087 CAS Number: 14283-07-9 EINECS: 238-178-9 UN: 3260 ADR: 8 II |
Specification Test Results
Appearance | White to gray Crystalline powder or crystals |
Elemental Analysis | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...