Ethylbenzene, anhydrous 99.8%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylethane

AC43380-0010 AC43380-1000
TS43380-0010EA 131 USD
TS43380-0010 TS43380-1000
Ethylbenzene, anhydrous 99.8%, AcroSeal™
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.86 g/cm³
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
UN: 1175
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.75 %
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR