Chlorobenzene ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzene ChloridePhenylchlorideMonochlorobenzene

AC40449-0010 AC40449-0025
TS40449-0025EA 205 USD
TS40449-0025 TS40449-0010
Chlorobenzene ≥99.5% ACS
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 131 °C (760 mmHg)
Melting Pt: –45 °C
Density: 1.1075 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://pr.vwr.com/stibo/web/std.lang.all/08/56/750856.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5230 to 1.5250 (20°C, 589 nm)
GC ≥99.5 %
Color scale ≤30 APHA
Residue after evaporation ≤0.02 %
Titrable acid ≤0.004 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR