Silver benzoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzoic acid silver (I) salt

AC21077-0100 AC21077-0500
TS21077-0100EA 75 USD
TS21077-0100 TS21077-0500
Silver benzoate 99%
Silver benzoate
Formula: C₇H₅AgO₂
MW: 228.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013030
CAS Number: 532-31-0
EINECS: 208-533-2
Merck Index: 13,01092

Order Now

Specification Test Results

Appearance White to beige to light gray Powder or crystals and/or chunks
Infrared spectrum Conforms
Silver (Ag) 46.4 to 47.8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR