Sodium benzoate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid sodium salt

AC14898-0010 AC14898-0050
TS14898-0010EA 47 USD
TS14898-0010 TS14898-0050
Sodium benzoate 99+%, Extra Pure
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or granules or flakes
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
Acidity/alkalinity ≤0.2 ml 0.1 M NaOH or HCl per gram
Titration with HClO4 ≥99.0 % (On dried substance)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR