Allyl bromide 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromopropene, 3-Propenyl bromide
AC10290-0025
AC10290-0050
AC10290-0100
AC10290-2500
AC10290-5000
TS10290-0025EA
314
USD
TS10290-0025
TS10290-0050
TS10290-0100
TS10290-2500
TS10290-5000
Allyl bromide 99% stabilised
Allyl bromide
Formula:
C₃H₅Br MW: 120.98 g/mol Boiling Pt: 70 Melting Pt: -119 Density: 1.39 g/cm³ Flash Pt: –1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000244 CAS Number: 106-95-6 EINECS: 203-446-6 UN: 1099 ADR: 3 I Merck Index: 13,00285 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4680 to 1.4700 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | 300 ppm propylene oxide |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...