Triiodomethane 99+%

Supplier: Thermo Scientific Chemicals
Warning

AC14912-0025 AC14912-1000 AC14912-5000
TS14912-0025EA 961 USD
TS14912-0025 TS14912-1000 TS14912-5000
Triiodomethane 99+%
Triiodomethane
Formula: CHI₃
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Merck Index: 13,05055

Order Now

Specification Test Results

Appearance Bright yellow to dark yellow Crystalline powder
Infrared spectrum Conforms
Melting point 119 to 122 °C
Loss on drying ≤0.5 % (24 h) (Over silicagel)
Residue after ignition ≤0.1 %
Titration ≥99.0 %
Chloride (Cl) ≤110 ppm
Sulfate (SO4) ≤170 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR