Lead tetraacetate 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

AC18062-0025 AC18062-1000 AC18062-5000
TS18062-0025EA 1054 USD
TS18062-0025 TS18062-1000 TS18062-5000
Lead tetraacetate 95% stabilized
Lead tetraacetate
Formula: (H₃CCOO)₄Pb
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008693
CAS Number: 546-67-8
UN: 1616
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to pink-orange Crystalline powder
Titration Iodometric ≥94 %
Stabilizer ca. 4 to 5 % Acetic acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR