Thallium(I) acetate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid thallium (I) salt, Thallous acetate

AC42077-0250 AC42077-1000 AC42077-5000
TS42077-0250EA 142 USD
TS42077-0250 TS42077-1000 TS42077-5000
Thallium(I) acetate 99%
Thallium(I) acetate
Formula: C₂H₃O₂Tl
MW: 263.43 g/mol
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013045
CAS Number: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR