Potassium acetate 99+% for analysis
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid potassium salt
AC21710-0010
AC21710-0050
AC21710-5000
TS21710-0010EA
95
USD
TS21710-0010
TS21710-0050
TS21710-5000
Potassium acetate 99+% for analysis
Potassium acetate
Formula:
CH₃COOK MW: 98.14 g/mol Melting Pt: 292 °C Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 EINECS: 204-822-2 Merck Index: 13,07687 |
Specification Test Results
Appearance | White Granular powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % |
pH | 7.5 to 9.0 (5% soln. at 20°C) |
Titration with HClO4 | 99.0 to 101.0 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...