Samarium trinitrate hexahydrate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Samarium (III) nitrate hexahydrate, Samarium(III) nitrate hydrate

AC31615-0250 AC31615-1000
TS31615-0250EA 90 USD
TS31615-0250 TS31615-1000
Samarium trinitrate hexahydrate 99.9% (trace metals basis)
Samarium trinitrate hexahydrate
Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Total conc. of imp.as REO ≤0.1 %
Samarium (Sm) 33.16 to 34.54 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR