Lanthanum(III) nitrate hexahydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21871-1000
AC21871-5000
TS21871-1000EA
145
USD
TS21871-1000
TS21871-5000
Lanthanum(III) nitrate hexahydrate 98+%
Lanthanum(III) nitrate hexahydrate
Formula:
La(NO₃)₃·6H₂O MW: 433.01 g/mol Boiling Pt: 126 Melting Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1 II Merck Index: 14,05363 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...