Cerium trinitrate hexahydrate 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

AC21869-0025 AC21869-1000 AC21869-5000
TS21869-0025EA 1031 USD
TS21869-0025 TS21869-1000 TS21869-5000
Cerium trinitrate hexahydrate 99.5%
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200
Melting Pt: 39
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1 III

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals and/or chunks
Assay ≥99.4 % (CeO2/TREO basis)
Sulfate (SO4) ≤200 ppm
Calcium oxide (CaO) ≤50 ppm
Iron(III)oxide (Fe2O3) ≤35 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR