Magnesium nitrate hexahydrate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals

AC19342-0100 AC19342-0500
TS19342-0100EA 55 USD
TS19342-0100 TS19342-0500
Magnesium nitrate hexahydrate 99.999% (trace metals basis)
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Boiling Pt: 330 °C (760 mmHg)
Melting Pt: 89 °C
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 9.1 to 9.7 % (Mg)
Total trace metal impurities ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR