Magnesium nitrate hexahydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals

AC21756-0010 AC21756-5000
TS21756-0010EA 137 USD
TS21756-0010 TS21756-5000
Magnesium nitrate hexahydrate 99+% for analysis
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Boiling Pt: 330
Melting Pt: 89
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
Merck Index: 13,05697

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric ≥99.0 %
pH 5.0 to 8.2 (5 % in water at 20°C)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Barium (Ba) ≤20 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤5 ppm
Strontium (Sr) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR