Sodium hydrogen sulphate monohydrate 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium bisulphate monohydrate

AC12500-0025 AC12500-0100 AC12500-5000
TS12500-0025EA 195 USD
TS12500-0025 TS12500-0100 TS12500-5000
Sodium hydrogen sulphate monohydrate 99% for analysis
Sodium hydrogen sulphate monohydrate
Formula: NaHSO₄·H₂O
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5

Order Now

Specification Test Results

Appearance White Adhering crystals
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.0005 %
Nitrate (NO3) ≤0.001 %
Phosphate (PO4) ≤0.001 %
Silicate (as SiO2) ≤0.005 %
Ammonium (NH4) ≤0.001 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤5 ppm
NH4OH precipitate ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR