Sodium hydrogen sulphate monohydrate 97%, crystals, extra pure reagent
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium bisulphate monohydrate
AC41943-0025
AC41943-0100
AC41943-5000
TS41943-0025EA
351
USD
TS41943-0025
TS41943-0100
TS41943-5000
Sodium hydrogen sulphate monohydrate 97%, crystals, extra pure reagent
Sodium hydrogen sulphate monohydrate
Formula:
NaHSO₄·H₂O MW: 138.08 g/mol Melting Pt: 183 Storage Temperature: Ambient |
MDL Number:
MFCD00149210 CAS Number: 10034-88-5 EINECS: 231-665-7 Merck Index: 13,08658 |
Specification Test Results
Appearance | White Adhering crystals |
Titration with NaOH | ≥96.0 % |
Chloride (Cl) | ≤20 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...