Lithium sulfate monohydrate 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC21833-0010 AC21833-1000
TS21833-0010EA 223 USD
TS21833-0010 TS21833-1000
Lithium sulfate monohydrate 99+%, pure
Lithium sulphate monohydrate
Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Melting Pt: 860 °C
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration after Ion exchange ≥99.0 % (On dry substance)
Loss on ignition 13.5 % to 15.0 % (600°C, 2 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR