Lithium hydroxide monohydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC41332-0025
AC41332-1000
AC41332-5000
TS41332-0025EA
931
USD
TS41332-0025
TS41332-1000
TS41332-5000
Lithium hydroxide monohydrate 98+% ACS
Lithium hydroxide monohydrate
Formula:
LiOH·1H₂O MW: 41.96 g/mol Boiling Pt: 920 Melting Pt: 462 Storage Temperature: Ambient |
MDL Number:
MFCD00149772 CAS Number: 1310-66-3 UN: 2680 ADR: 8 II Merck Index: 13,05556 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration with HCl | ≥98.0 % |
Chloride (Cl) | ≤0.01 % |
Sulfate (SO4) | ≤0.05 % |
Lithium carbonate | ≤2.0 % |
Iron (Fe) | ≤0.002 % |
Heavy metals (ICP-OES) | ≤0.002 % |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...