Sodium perchlorate monohydrate 85+% ACS

Supplier: Thermo Scientific Chemicals
Danger

AC34363-0010 AC34363-0025 AC34363-1000
TS34363-0010EA 287 USD
TS34363-0010 TS34363-0025 TS34363-1000
Sodium perchlorate monohydrate 85+% ACS
Sodium perchlorate monohydrate
Formula: NaClO₄·H₂O
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00149164
CAS Number: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798

Order Now

Specification Test Results

Appearance White Adhering crystals
Titration Argentometric 85.0 to 90.0 % (as NaClO4)
pH 6.0 to 8.0 (5 % solution at 25.0°C)
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤20 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤500 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR