Sodium carbonate decahydrate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Soda

AC27107-0025 AC27107-0100 AC27107-1000
TS27107-0025EA 83 USD
TS27107-0025 TS27107-0100 TS27107-1000
Sodium carbonate decahydrate 99+%, Extra Pure
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
MW: 286.14 g/mol
Melting Pt: 34 °C
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Appearance White Crystals
Titration with HCl ≥99.0 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Aluminium (Al) ≤10 ppm
Iron (Fe) ≤4 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤50 ppm
Silicon dioxide (SiO2) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR