Sodium carbonate decahydrate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Soda
AC21350-0010
AC21350-0025
TS21350-0010EA
85
USD
TS21350-0010
TS21350-0025
Sodium carbonate decahydrate 99+% for analysis
Sodium carbonate decahydrate
Formula:
Na₂CO₃·10H₂O MW: 286.14 g/mol Melting Pt: 34 °C Density: 1.46 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149178 CAS Number: 6132-02-1 EINECS: 207-838-8 |
Specification Test Results
Appearance | White Crystals |
Titration with HCl | ≥99.0 % |
Chloride (Cl) | ≤5 ppm |
PO4 and SiO2 (as SiO2) | ≤0.002 % |
Aluminium (Al) | ≤5 ppm |
Calcium (Ca) | ≤20 ppm |
Iron (Fe) | ≤2 ppm |
Magnesium (Mg) | ≤2 ppm |
Potassium (K) | ≤100 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Total nitrogen (as N) | ≤0.0005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...