Lead(II) acetate trihydrate 99.999% (trace metals basis), extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

AC31723-0050 AC31723-0250
TS31723-0050EA 36 USD
TS31723-0050 TS31723-0250
Lead(II) acetate trihydrate 99.999% (trace metals basis), extra pure
Lead(II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280
Melting Pt: 75
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR