Brady® Self-Laminating Vinyl Wrap-Around Labels with Ribbon, Brady
Supplier: Brady Worldwide
M21-11-427 M21-18-427
76537-146RL 37.36 USD
76537-146 76537-148
Brady® Self-Laminating Vinyl Wrap-Around Labels with Ribbon, Brady
Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear self-laminating protective layer. Print almost anywhere with a durable Brady portable label printer.
  • Permanent acrylic adhesive
  • Matte finish
  • Flame-retardant, self-laminating labels
Designed for use on tightly curved surfaces such as wires and cables. Self-extinguishing and resistant to abrasion, solvents, oil, grease, weather, and extreme temperatures. Made with a permanent acrylic adhesive that works great on smooth, rough, powder-coated, and irregular surfaces. A clear, non-printable overlaminate provides increased protection. For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
Certificaciones: UL recognized to UL 2238; RoHS compliant to 2011/65/EU (Pass 2015/863).