Anti-ACE2 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Supplier: Sino Biological
10108MM36P100 10108MM36P25
76535-506EA 202 USD
76535-506 76535-510
Anti-ACE2 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog#10108-H08H; NP_068576.1; Met1-Ser740) and conjugated with PE under optimum conditions, the unreacted PE was removed.