Methyl mercaptan sodium salt 90+%, technical

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt

AC46989-0010 AC46989-0250 AC46989-1000
TS46989-0010EA 41 USD
TS46989-0010 TS46989-0250 TS46989-1000
Methyl mercaptan sodium salt 90+%, technical
Methyl mercaptan sodium salt
Formula: CH₃NaS
MW: 70.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174316
CAS Number: 5188-07-8
UN: 2921
ADR: 8,I

Order Now

Specification Test Results

Assay 90+%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR