VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Supplier: Avantor
GeneMate
H3770-HS-GM H3770-HS-P1-GM
76538-828EA 602.38 USD
76538-828 76538-822
VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
GeneMate™ Magnetic Hotplate Stirrers, 7×7" with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • Excellent chemical resistance with white ceramic work surface
  • Stir volumes up to 5 L
  • Safety LED warns user when top plate is unsafe to touch
  • Choice of model with temperature probe for control over actual sample temperature
Certificaciones: CE marked.
Información para pedidos: 76538-828 is supplied with external temperature probe for control over actual sample temperature. Full specification details of stirrer and hotplate only models are given under separate entries, please view full details by searching for the relevant product code.