Acetal 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,1-Diethoxyethane, Acetaldehyde diethyl acetal
10222-2500
10222-0010
10222-0050
TS10222-2500EA
100
USD
TS10222-2500
TS10222-0010
TS10222-0050
Acetal 99%
Acetal
Formula:
C₆H₁₄O₂ Boiling Pt: 102.7 °C (1013 hPa) Melting Pt: –100 °C Density: 0.825 g/cm³ Flash Pt: –21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009243 CAS Number: 105-57-7 EINECS: 203-310-6 UN: 1088 ADR: 3,II Merck Index: 13,00040 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3790 to 1.3850 (20°C, 589 nm) |
Density | (20°C) 0.822 to 0.829 kg/l |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...