CytoWatch™ QZ100 Monocyte Blocking Reagent, AAT Bioquest
Supplier: AAT BIOQUEST INC
37000 37001
76485-716EA 90.09 USD
76485-716 76485-718
CytoWatch™ QZ100 Monocyte Blocking Reagent, AAT Bioquest
Assays Cellular Assays
Some dye-labeled fluorescent antibody conjugates used in cell surface staining of live cells often exhibit non-specific binding in monocytes and macrophages.
  • Suitable for PBMC or whole blood
  • Minimizes background signal
  • No effect on lymphocytes, monocytes, or granulocytes
Precaución: For research use only.