Screen Quest™ Calbryte-590 Probenecid-Free and Wash-Free Calcium Assay Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
36200 36201 36202
76484-412EA 375.27 USD
76484-412 76484-414 76484-416
Screen Quest™ Calbryte-590 Probenecid-Free and Wash-Free Calcium Assay Kit, AAT Bioquest
Assays Cellular Assays
Calcium flux assays are the preferred methods for screening G protein coupled receptors (GPCRs) in drug discovery.
  • 100 to 250 times fluorescence increase once bound to calcium
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photostable signal
Precaución: For research use only.