Protein A-Agarose Resin
Supplier: AAT BIOQUEST INC
55000 55005
76484-340EA 124.31 USD
76484-340 76484-342
Protein A-Agarose Resin
Proteins and Peptides
Protein A agarose was prepared by covalently immobilizing protein A onto high-quality crosslinked beaded agarose.
  • High affinity
  • Targets igg antibodies
  • pH stable between 3 to 13
Precaución: For research use only.