Anti-IgG Goat Polyclonal Antibody (iFluor® 532)
Supplier: AAT BIOQUEST INC
16536 16775
76482-494EA 124.31 USD
76482-494 76482-496
Anti-IgG Goat Polyclonal Antibody (iFluor® 532)
Antibodies
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Precaución: For research use only.