MagaDye™ 535-ddGTP, AAT Bioquest
Supplier: AAT BIOQUEST INC
17063 17067
76485-284EA 1142.29 USD
76485-284 76485-286
MagaDye™ 535-ddGTP, AAT Bioquest
Nucleic Acid Reagents Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Precaución: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=888fb87da42acc1a75ccc90ed26654142ace160e&vid=3dd7a3988de6808deee0d738f71272d0af5722e7&prid=web&clid=SAPDAM
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more