Cal-520FF™ AM
Supplier: AAT BIOQUEST INC
|
Warning
|
21142
21143
76483-296EA
626.51
USD
76483-296
76483-298
Cal-520FF™ AM
Cal-520FF™ AM
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell permeant
Precaución: For research use only
MW:
1138.92 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...