Cal-520® potassium salt

Supplier: AAT BIOQUEST INC
Warning

21140 21141
76483-292EA 375.27 USD
76483-292 76483-294
Cal-520® potassium salt
Cal-520® potassium salt

Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.


  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell impermeant


Precaución: For research use only

MW: 921.08 g/mol
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR