Cal-520® sodium salt
Supplier: AAT BIOQUEST INC
|
Warning
|
21135
21136
76483-288EA
375.27
USD
76483-288
76483-290
Cal-520® sodium salt
Cal-520® sodium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell impermeant
Precaución: For research use only
MW:
840.54 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...