Cal-520® AM

Supplier: AAT BIOQUEST INC
Warning

21130 21131
76483-284EA 249.58 USD
76483-284 76483-286
Cal-520® AM
Cal-520® AM

Cal-520® AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.


  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant


Precaución: For research use only

MW: 1102.95 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR