Rhod-4™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

21119 21129
76483-272EA 626.51 USD
76483-272 76483-274
Rhod-4™ potassium salt
Rhod-4™ potassium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Long-wavelength emission
  • Cell impermeant


Precaución: For research use only

MW: 880.07 g/mol
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR