Fura-10™ AM
Supplier: AAT BIOQUEST INC
Warning
21114 21115
76485-318EA 117.59 USD
76485-318 76485-320
Fura-10™ AM
Fura-10™ AM
Among ratiometric calcium ion indicators, Fura-2 and Indo-1 are the two most popular ones.
  • Calcium-dependent fluorescence
  • Ratiometric detection
  • Improved cell retention
  • Cell permeant
Precaución: For research use only
ADR: 5,II