Fluo-8FF™ AM
Supplier: AAT BIOQUEST INC
Warning
21104 21105
76483-264EA 249.58 USD
76483-264 76483-266
Fluo-8FF™ AM
Fluo-8FF™ AM
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms
Precaución: For research use only
ADR: 5,II