Fluo-8FF™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

21102 21103
76483-260EA 249.58 USD
76483-260 76483-262
Fluo-8FF™ potassium salt
Fluo-8FF™ potassium salt

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms


Precaución: For research use only

MW: 913.05 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b8ca1c7d2adf6e493367f128bf02eab61bb3ea4&vid=10720b497f99bfcd321ee5b7ff04bce8b92f46a4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR