Fluo-8L™ potassium salt

Supplier: AAT BIOQUEST INC
Warning

21100 21101
76483-256EA 249.58 USD
76483-256 76483-258
Fluo-8L™ potassium salt
Fluo-8L™ potassium salt

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms


Precaución: For research use only

MW: 909.09 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR