Fluo-8L™ sodium salt

Supplier: AAT BIOQUEST INC
Warning

21098 21099
76483-252EA 249.58 USD
76483-252 76483-254
Fluo-8L™ sodium salt
Fluo-8L™ sodium salt

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms


Precaución: For research use only

MW: 828.54 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR