Fluo-8L™ AM
Supplier: AAT BIOQUEST INC
|
Warning
|
21096
21097
76483-248EA
375.27
USD
76483-248
76483-250
Fluo-8L™ AM
Fluo-8L™ AM
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Precaución: For research use only
MW:
1078.95 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...