Fluo-8H™ AM

Supplier: AAT BIOQUEST INC
Warning

21090 21091
76483-242EA 375.27 USD
76483-242 76483-244
Fluo-8H™ AM
Fluo-8H™ AM

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms


Precaución: For research use only

MW: 1074.98 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR