Fluo-8® sodium salt

Supplier: AAT BIOQUEST INC
Warning

21086 21088
76483-234EA 375.27 USD
76483-234 76483-236
Fluo-8® sodium salt
Fluo-8® sodium salt

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms


Precaución: For research use only

MW: 796.53 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a3f72d5f07fde60218aad0be63d5a249b2dbf2&vid=7a3ef5f1761f8e754b0ced2488c6354445e760b4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR