Fluo-8 AM
Supplier: AAT BIOQUEST INC
|
Warning
|
21080
21081
21082
21083
76483-226EA
223.27
USD
76483-226
76483-228
76483-230
76483-232
Fluo-8 AM
Fluo-8 AM
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Precaución: For research use only
MW:
1046.93 g/mol |
CAS Number:
1345980-40-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...