Rhod-FF AM
Supplier: AAT BIOQUEST INC
|
Warning
|
21077
21078
76483-222EA
375.27
USD
76483-222
76483-224
Rhod-FF AM
Rhod-FF AM
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Low binding affinity
- Cell permeant
Precaución: For research use only
MW:
1145.9 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...