Rhod-FF tripotassium salt
Supplier: AAT BIOQUEST INC
|
Warning
|
21075
21076
76483-218EA
375.27
USD
76483-218
76483-220
Rhod-FF tripotassium salt
Rhod-FF tripotassium salt
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Low binding affinity
- Cell impermeant
Precaución: For research use only
ADR:
5,II |