Fura-8™ AM
Supplier: AAT BIOQUEST INC
Warning
21055 21056
76483-194EA 124.31 USD
76483-194 76483-196
Fura-8™ AM
Fura-8™ AM
Although Fura-2 has become the preferred excitation-ratioable calcium indicator of choice, it has certain limitations (e.g., lower sensitivity than single wavelength calcium indicators such as Fluo-8® and Cal-520®).
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell permeant
Precaución: For research use only
ADR: 5,II